Assay Details
| Gene Symbol: NMS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626008.2 | XP_005626065.1 | 3 - 4 | 784 | 90 | |
| XM_014117466.2 | XP_013972941.1 | 2 - 3 | 2662 | 90 | ||
| XM_022424581.1 | XP_022280289.1 | 3 - 4 | 2098 | 90 | ||
| XM_022424582.1 | XP_022280290.1 | 3 - 4 | 1479 | 90 | ||
| XM_022424583.1 | XP_022280291.1 | 2 - 3 | 2666 | 90 | ||
| XM_549401.5 | XP_549401.3 | 2 - 3 | 614 | 90 | ||
More Information
Set Membership: |
|

NMS
Genomic Map