Assay Details
| Gene Symbol: NAGK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022404855.1 | XP_022260563.1 | 2 - 3 | 449 | 86 | |
| XM_022404856.1 | XP_022260564.1 | 2 - 3 | 452 | 86 | ||
| XM_549180.7 | XP_549180.3 | 2 - 3 | 253 | 86 | ||
More Information
Set Membership: |
|

NAGK
Genomic Map