Assay Details
Gene Symbol: RPP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252381.1 | NP_001239310.1 | 1 - 2 | 67 | 76 | |
XM_005627042.3 | XP_005627099.2 | 1 - 2 | 136 | 76 | ||
GenBank mRNA | DN370541.1 | - | 1 - 2 | 67 | 76 | |
DN378554.1 | - | 1 - 2 | 42 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
