Assay Details
| Gene Symbol: SCML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014111799.2 | XP_013967274.2 | 2 - 3 | 415 | 121 | |
| XM_014111803.2 | XP_013967278.1 | 2 - 3 | 413 | 121 | ||
| XM_014111804.2 | XP_013967279.1 | 2 - 3 | 207 | 121 | ||
| XM_014111805.2 | XP_013967280.1 | 2 - 3 | 289 | 121 | ||
| XM_022415760.1 | XP_022271468.1 | 2 - 3 | 246 | 121 | ||
| XM_022415763.1 | XP_022271471.1 | 2 - 3 | 412 | 121 | ||
| XM_022415764.1 | XP_022271472.1 | 2 - 3 | 413 | 121 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014111799.2, XM_014111803.2, XM_014111804.2, XM_014111805.2, XM_022415760.1, XM_022415763.1, XM_022415764.1 transcripts.
Set Membership: |
|

SCML1
Genomic Map