Assay Details
| Gene Symbol: ZNF248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_843150.5 | XP_848243.1 | 535 | 137 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_843150.5 transcripts.
Set Membership: |
|

ZNF248
Genomic Map