Assay Details
| Gene Symbol: GDPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624747.3 | XP_005624804.1 | 5 - 6 | 461 | 72 | |
| XM_548237.6 | XP_548237.3 | 5 - 6 | 614 | 72 | ||
More Information
Set Membership: |
|

GDPD1
Genomic Map