Assay Details
| Gene Symbol: LZIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639059.4 | XP_003639107.2 | 1 | 242 | 106 | |
| XM_014113920.2 | XP_013969395.1 | 375 | 106 | |||
| XM_014113921.2 | XP_013969396.1 | 426 | 106 | |||
| XM_014113922.2 | XP_013969397.1 | 375 | 106 | |||
| XM_014113923.2 | XP_013969398.1 | 1 | 255 | 106 | ||
| XM_022419035.1 | XP_022274743.1 | 375 | 106 | |||
| XM_022419036.1 | XP_022274744.1 | 830 | 106 | |||
| XM_022419037.1 | XP_022274745.1 | 1 | 230 | 106 | ||
| XM_022419038.1 | XP_022274746.1 | 777 | 106 | |||
More Information
Set Membership: |
|

LZIC
Genomic Map