Assay Details
Gene Symbol: CYP4V2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022404181.1 | XP_022259889.1 | 4 - 5 | 451 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
