Assay Details
| Gene Symbol: LOC111091303 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022406528.1 | XP_022262236.1 | 2 - 3 | 143 | 77 | |
| Gene Symbol: RPP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_844095.5 | XP_849188.2 | 4 - 5 | 368 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022406528.1 transcripts.
Set Membership: |
|

LOC111091303
Genomic Map