Assay Details
Gene Symbol: LOC606786 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014113267.2 | XP_013968742.1 | 940 | 55 | ||
XM_022418064.1 | XP_022273772.1 | 928 | 55 | |||
XM_022418065.1 | XP_022273773.1 | 1206 | 55 | |||
XM_022418066.1 | XP_022273774.1 | 830 | 55 | |||
XM_843133.4 | XP_848226.1 | 939 | 55 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
