Assay Details
Gene Symbol: AURKAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620384.3 | XP_005620441.1 | 747 | 76 | ||
XM_014113810.2 | XP_013969285.1 | 740 | 76 | |||
XM_843641.5 | XP_848734.1 | 733 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
