Assay Details
Gene Symbol: BFSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005634760.3 | XP_005634817.1 | 3 - 4 | 539 | 64 | |
XM_005634761.3 | XP_005634818.1 | 4 - 5 | 378 | 64 | ||
XM_005634762.2 | XP_005634819.1 | 4 - 5 | 393 | 64 | ||
XM_022409397.1 | XP_022265105.1 | 4 - 5 | 307 | 64 | ||
XM_022409398.1 | XP_022265106.1 | 5 - 6 | 506 | 64 | ||
XM_022409399.1 | XP_022265107.1 | 5 - 6 | 480 | 64 | ||
XM_022409400.1 | XP_022265108.1 | 5 - 6 | 490 | 64 | ||
XM_022409401.1 | XP_022265109.1 | 5 - 6 | 510 | 64 | ||
XM_843800.5 | XP_848893.1 | 3 - 4 | 539 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
