Assay Details
Gene Symbol: SLIRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_547932.6 | XP_547932.2 | 2 - 3 | 216 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
