Assay Details
Gene Symbol: LOC609830 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638865.3 | XP_005638922.1 | 3 - 4 | 440 | 125 | |
XM_847173.5 | XP_852266.1 | 3 - 4 | 467 | 125 |
Gene Symbol: IFNAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003640073.4 | XP_003640121.1 | 3 - 4 | 467 | 125 | |
XM_005638840.3 | XP_005638897.1 | 3 - 4 | 440 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
