Assay Details
| Gene Symbol: IL21R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621391.3 | XP_005621448.1 | 5 - 6 | 1100 | 67 | |
| XM_844902.4 | XP_849995.1 | 4 - 5 | 641 | 67 | ||
More Information
Set Membership: |
|

IL21R
Genomic Map