Assay Details
| Gene Symbol: THEM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014120713.2 | XP_013976188.1 | 3 - 4 | 471 | 102 | |
| XM_022405013.1 | XP_022260721.1 | 3 - 4 | 471 | 102 | ||
| XM_846372.5 | XP_851465.2 | 3 - 4 | 471 | 102 | ||
More Information
Set Membership: |
|

THEM4
Genomic Map