Assay Details
Gene Symbol: UPF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005634141.2 | XP_005634198.1 | 2 - 3 | 352 | 138 | |
XM_022408201.1 | XP_022263909.1 | 2 - 3 | 352 | 138 | ||
XM_022408202.1 | XP_022263910.1 | 2 - 3 | 352 | 138 | ||
XM_022408203.1 | XP_022263911.1 | 2 - 3 | 352 | 138 | ||
XM_022408204.1 | XP_022263912.1 | 2 - 3 | 180 | 138 | ||
XM_022408205.1 | XP_022263913.1 | 2 - 3 | 359 | 138 | ||
XM_022408206.1 | XP_022263914.1 | 2 - 3 | 360 | 138 | ||
XM_022408208.1 | XP_022263916.1 | 2 - 3 | 359 | 138 | ||
XM_022408209.1 | XP_022263917.1 | 2 - 3 | 360 | 138 | ||
XM_022408210.1 | XP_022263918.1 | 2 - 3 | 360 | 138 | ||
XM_022408211.1 | XP_022263919.1 | 2 - 3 | 360 | 138 | ||
XM_022408212.1 | XP_022263920.1 | 2 - 3 | 360 | 138 | ||
XM_022408213.1 | XP_022263921.1 | 2 - 3 | 360 | 138 | ||
XM_844659.5 | XP_849752.2 | 2 - 3 | 352 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
