Assay Details
Gene Symbol: CMTM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014106494.2 | XP_013961969.1 | 3 - 4 | 394 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
