Assay Details
Gene Symbol: HPCAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630116.3 | XP_005630173.1 | 600 | 61 | ||
XM_014120504.2 | XP_013975979.1 | 502 | 61 | |||
XM_847124.3 | XP_852217.2 | 657 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
