Assay Details
Gene Symbol: ZNF800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005628427.3 | XP_005628484.1 | 4 - 5 | 484 | 123 | |
XM_005628429.2 | XP_005628486.1 | 4 - 5 | 496 | 123 | ||
XM_005628430.3 | XP_005628487.1 | 2 - 3 | 280 | 123 | ||
XM_022427269.1 | XP_022282977.1 | 3 - 4 | 400 | 123 | ||
XM_022427270.1 | XP_022282978.1 | 3 - 4 | 388 | 123 | ||
XM_532439.6 | XP_532439.4 | 4 - 5 | 496 | 123 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005628430.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
