Assay Details
Gene Symbol: ITGB3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620209.3 | XP_005620266.1 | 4 - 5 | 410 | 101 | |
XM_005620210.3 | XP_005620267.1 | 3 - 4 | 336 | 101 | ||
XM_005620211.3 | XP_005620268.1 | 3 - 4 | 333 | 101 | ||
XM_014113718.2 | XP_013969193.1 | 4 - 5 | 409 | 101 | ||
XM_022418761.1 | XP_022274469.1 | 4 - 5 | 408 | 101 | ||
XM_022418762.1 | XP_022274470.1 | 4 - 5 | 409 | 101 | ||
XM_022418763.1 | XP_022274471.1 | 4 - 5 | 409 | 101 | ||
XM_022418764.1 | XP_022274472.1 | 4 - 5 | 401 | 101 | ||
XM_022418765.1 | XP_022274473.1 | 4 - 5 | 411 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
