Assay Details
Gene Symbol: PUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005636079.3 | XP_005636136.1 | 614 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
