Assay Details
Gene Symbol: MED9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_846808.4 | XP_851901.1 | 1 - 2 | 311 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Molecular Function -



