Assay Details
| Gene Symbol: ERI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621466.3 | XP_005621523.1 | 4 - 5 | 481 | 103 | |
| XM_022420004.1 | XP_022275712.1 | 5 - 6 | 627 | 103 | ||
| XM_022420005.1 | XP_022275713.1 | 4 - 5 | 544 | 103 | ||
| XM_022420006.1 | XP_022275714.1 | 5 - 6 | 561 | 103 | ||
| XM_022420007.1 | XP_022275715.1 | 4 - 5 | 478 | 103 | ||
| XM_846013.5 | XP_851106.2 | 5 - 6 | 565 | 103 | ||
More Information
Set Membership: |
|

ERI2
Genomic Map