Assay Details
| Gene Symbol: SLCO3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003434373.4 | XP_003434421.1 | 3 - 4 | 750 | 101 | |
| XM_845970.5 | XP_851063.2 | 3 - 4 | 750 | 101 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLCO3A1
Genomic Map