Assay Details
Gene Symbol: ANKMY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639483.4 | XP_003639531.1 | 3 - 4 | 465 | 68 | |
XM_005628583.3 | XP_005628640.1 | 3 - 4 | 219 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
