Assay Details
| Gene Symbol: LOC609365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622321.3 | XP_005622378.1 | 499 | 135 | ||
| XM_005622322.3 | XP_005622379.1 | 499 | 135 | |||
| XM_005622323.3 | XP_005622380.1 | 499 | 135 | |||
| XM_005622324.3 | XP_005622381.1 | 499 | 135 | |||
| XM_005622325.3 | XP_005622382.1 | 499 | 135 | |||
| XM_005622326.3 | XP_005622383.1 | 499 | 135 | |||
| XM_022420857.1 | XP_022276565.1 | 499 | 135 | |||
More Information
Set Membership: |
|

LOC609365
Genomic Map