Assay Details
Gene Symbol: TEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_846734.5 | XP_851827.1 | 144 | 118 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_846734.5 transcripts.
Set Membership: |
![]() ![]() |
