Assay Details
Gene Symbol: TEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624079.2 | XP_005624136.1 | 258 | 81 | ||
XM_005624080.3 | XP_005624137.1 | 216 | 81 | |||
XM_022422716.1 | XP_022278424.1 | 1058 | 81 | |||
XM_846734.5 | XP_851827.1 | 267 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
