Assay Details
| Gene Symbol: POLR3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618122.3 | XP_005618179.1 | 302 | 103 | ||
| XM_005618123.3 | XP_005618180.1 | 302 | 103 | |||
| XM_014112225.2 | XP_013967700.1 | 174 | 103 | |||
| XM_014112227.2 | XP_013967702.1 | 174 | 103 | |||
| XM_022416728.1 | XP_022272436.1 | 302 | 103 | |||
| XM_022416729.1 | XP_022272437.1 | 302 | 103 | |||
| XM_847057.5 | XP_852150.1 | 262 | 103 | |||
More Information
Set Membership: |
|

POLR3G
Genomic Map