Assay Details
Gene Symbol: ARPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252380.1 | NP_001239309.1 | 1 - 2 | 157 | 117 | |
GenBank mRNA | DN434974.1 | - | 1 - 2 | 166 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() |
