Assay Details
Gene Symbol: ARPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252380.1 | NP_001239309.1 | 4 - 5 | 570 | 98 | |
XM_005618244.1 | XP_005618301.1 | 4 - 5 | 881 | 98 | ||
XM_022415725.1 | XP_022271433.1 | 4 - 5 | 484 | 98 | ||
GenBank mRNA | DN369634.1 | - | 1 - 2 | 97 | 99 | |
DN434974.1 | - | 4 - 5 | 579 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
