Assay Details
| Gene Symbol: LOC608483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014108242.2 | XP_013963717.1 | 6 - 7 | 869 | 106 | |
| XM_022410822.1 | XP_022266530.1 | 5 - 6 | 1513 | 106 | ||
More Information
Set Membership: |
|

LOC608483
Genomic Map