Assay Details
| Gene Symbol: RSPH10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621110.3 | XP_005621167.1 | 3316 | 103 | ||
| XM_005621112.3 | XP_005621169.1 | 2228 | 103 | |||
| XM_005621113.3 | XP_005621170.1 | 2923 | 103 | |||
| XM_014114297.2 | XP_013969772.1 | 2158 | 103 | |||
| XM_022419724.1 | XP_022275432.1 | 3068 | 103 | |||
| XM_022419725.1 | XP_022275433.1 | 2999 | 103 | |||
| XM_846164.5 | XP_851257.1 | 3385 | 103 | |||
More Information
Set Membership: |
|

RSPH10B
Genomic Map