Assay Details
| Gene Symbol: B2M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284479.1 | NP_001271408.1 | 1 - 2 | 90 | 87 | |
| XM_003640047.2 | XP_003640095.1 | 1 - 2 | 294 | 87 | ||
| GenBank mRNA | JQ733515.1 | - | 1 - 2 | 90 | 87 | |
More Information
Set Membership: |
|

B2M
Genomic Map