Assay Details
| Gene Symbol: PLAC8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_845786.4 | XP_850879.1 | 1 - 2 | 109 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_845786.4 transcripts.
Set Membership: |
|

PLAC8A
Genomic Map