Assay Details
| Gene Symbol: PLAC8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022413579.1 | XP_022269287.1 | 3 - 4 | 523 | 76 | |
| XM_845786.4 | XP_850879.1 | 3 - 4 | 368 | 76 | ||
More Information
Set Membership: |
|

PLAC8A
Genomic Map