Assay Details
Gene Symbol: GEM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638130.3 | XP_005638187.1 | 3 - 4 | 669 | 62 | |
XM_014109322.2 | XP_013964797.1 | 3 - 4 | 868 | 62 | ||
XM_022412352.1 | XP_022268060.1 | 3 - 4 | 868 | 62 | ||
XM_849321.5 | XP_854414.2 | 3 - 4 | 860 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
