Assay Details
| Gene Symbol: KLRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637176.2 | XP_005637233.1 | 3 - 4 | 506 | 72 | |
| XM_014108512.2 | XP_013963987.1 | 3 - 4 | 844 | 72 | ||
| XM_022411316.1 | XP_022267024.1 | 3 - 4 | 182 | 72 | ||
| XM_022411317.1 | XP_022267025.1 | 3 - 4 | 955 | 72 | ||
| XM_022411319.1 | XP_022267027.1 | 4 - 5 | 904 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022411319.1 transcripts.
Set Membership: |
|

KLRG1
Genomic Map