Assay Details
| Gene Symbol: DMRTC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616507.3 | XP_005616564.3 | 6 - 7 | 866 | 91 | |
| XM_005616509.3 | XP_005616566.1 | 6 - 7 | 917 | 91 | ||
| XM_005616510.2 | XP_005616567.1 | 6 - 7 | 918 | 91 | ||
| XM_005616511.2 | XP_005616568.1 | 6 - 7 | 917 | 91 | ||
| XM_022403405.1 | XP_022259113.1 | 6 - 7 | 900 | 91 | ||
| XM_022403406.1 | XP_022259114.1 | 6 - 7 | 798 | 91 | ||
More Information
Set Membership: |
|

DMRTC2
Genomic Map