Assay Details
Gene Symbol: LSM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014119216.2 | XP_013974691.1 | 2 - 3 | 247 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
