Assay Details
| Gene Symbol: LOC102154691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625569.3 | XP_005625626.1 | 1 | 71 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005625569.3 transcripts.
Set Membership: |
|

LOC102154691
Genomic Map