Assay Details
Gene Symbol: SRPRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005634450.3 | XP_005634507.1 | 3 - 4 | 375 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
