Assay Details
| Gene Symbol: LOC610465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014114019.2 | XP_013969494.1 | 4 - 5 | 640 | 58 | |
| XM_014114020.2 | XP_013969495.1 | 4 - 5 | 638 | 58 | ||
| XM_014114021.2 | XP_013969496.1 | 3 - 4 | 587 | 58 | ||
More Information
Set Membership: |
|

LOC610465
Genomic Map