Assay Details
| Gene Symbol: PRRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003638818.3 | XP_003638866.1 | 4 - 5 | 473 | 67 | |
| XM_005616308.2 | XP_005616365.1 | 4 - 5 | 455 | 67 | ||
More Information
Set Membership: |
|

PRRG2
Genomic Map