Assay Details
| Gene Symbol: MAP2K2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048136.1 | NP_001041601.1 | 5 - 6 | 598 | 60 | |
| GenBank mRNA | DQ489531.1 | - | 5 - 6 | 598 | 60 | |
More Information
Set Membership: |
|

MAP2K2
Genomic Map