Assay Details
Gene Symbol: PAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638073.3 | XP_005638130.1 | 8 - 9 | 1358 | 76 | |
XM_005638074.3 | XP_005638131.1 | 8 - 9 | 1342 | 76 | ||
XM_005638075.3 | XP_005638132.1 | 7 - 8 | 1280 | 76 | ||
XM_005638077.3 | XP_005638134.1 | 7 - 8 | 1232 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
