Assay Details
| Gene Symbol: ACOT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003434192.4 | XP_003434240.1 | 3 - 4 | 1681 | 98 | |
| XM_022414625.1 | XP_022270333.1 | 3 - 4 | 1662 | 98 | ||
| XM_848449.5 | XP_853542.1 | 2 - 3 | 379 | 98 | ||
More Information
Set Membership: |
|

ACOT13
Genomic Map