Assay Details
Gene Symbol: SMPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022415964.1 | XP_022271672.1 | 2 - 3 | 133 | 64 | |
XM_022415965.1 | XP_022271673.1 | 2 - 3 | 192 | 64 | ||
XM_849474.5 | XP_854567.1 | 2 - 3 | 287 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
