Assay Details
| Gene Symbol: SELENOF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114760.2 | NP_001108232.1 | 3 - 4 | 589 | 141 | |
| GenBank mRNA | CX004807.1 | - | 3 - 4 | 273 | 141 | |
| DN394348.1 | - | 3 - 4 | 301 | 141 | ||
| DN417247.1 | - | 3 - 4 | 595 | 142 | ||
More Information
Set Membership: |
|

SELENOF
Genomic Map